Gene Transcripts

Gene Symbol
Mup20
Entrez Gene ID
381530
Gene Name
major urinary protein 20
Gene Aliases
Gm12560, Mup24, OTTMUSG00000007485, darcin
Chromosome Location
Chr. 4: 62011406 - 62054143 on Build GRCm38
UniGene ID
Mm.460005
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001012323.1 NP_001012323.1 2 187
GenBank mRNA BC089613.1 - 2 187
BC092096.1 - 2 213 4161847
EU882234.1 - 2 119

View Full Product Details