Gene Transcripts

Gene Symbol
Olfml1
Entrez Gene ID
361621
Gene Name
olfactomedin-like 1
Gene Aliases
Chromosome Location
Chr. 1: 171793782 - 171818074 on Build Rnor_6.0
UniGene ID
Rn.22729
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001013192.1 NP_001013210.1 3 2824
GenBank mRNA BC081972.1 - 3 2824
FQ213807.1 - 1 948

View Full Product Details