Gene Transcripts

Gene Symbol
Zdhhc12
Entrez Gene ID
366014
Gene Name
zinc finger, DHHC-type containing 12
Gene Aliases
Chromosome Location
Chr. 3: 8656397 - 8659102 on Build Rnor_6.0
UniGene ID
Rn.95138
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001013239.1 NP_001013257.1 5 1065
GenBank mRNA AY886528.1 - 5 1065
BC076393.1 - 5 1065 7107692

View Full Product Details