Gene Transcripts
- Gene Symbol
- HAUS4
- Entrez Gene ID
- 54930
- Gene Name
- HAUS augmin like complex subunit 4
- Gene Aliases
- C14orf94
- Chromosome Location
- Chr. 14: 22946228 - 22957142 on Build GRCh38
- UniGene ID
- Hs.442782
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NM_001166269.1 |
NP_001159741.1
|
3, 4 |
455 |
|
NM_001166270.1 |
NP_001159742.1
|
3, 4 |
447 |
|
NM_017815.2 |
NP_060285.2
|
3, 4 |
447 |
|
GenBank mRNA |
AK000431.1 |
- |
3, 4 |
416 |
|
AK026016.1 |
- |
3, 4 |
401 |
|
AK057779.1 |
- |
3, 4 |
523 |
|
BC001916.1 |
- |
3, 4 |
444 |
2905474 |
BC002554.2 |
- |
3, 4 |
378 |
3139198 |
BQ066232.1 |
- |
3, 4 |
455 |
|
BX247981.1 |
- |
3, 4 |
631 |
|
BX248256.1 |
- |
3, 4 |
414 |
|
View Full Product Details