Gene Transcripts

Gene Symbol
Mtg2
Entrez Gene ID
296462
Gene Name
mitochondrial ribosome-associated GTPase 2
Gene Aliases
Gtpbp5
Chromosome Location
Chr. 3: 175448313 - 175464109 on Build Rnor_6.0
UniGene ID
Rn.17330
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001013924.1 NP_001013946.1 7 972
XM_006235817.2 XP_006235879.1 7 1398
XM_006235818.2 XP_006235880.1 7 1018
XM_006235820.2 XP_006235882.1 7 1026
XM_008762485.1 XP_008760707.1 6 2532
XM_008762486.1 XP_008760708.1 7 1402
XM_008762487.1 XP_008760709.1 8 1119
XM_008762488.1 XP_008760710.1 8 1142
XM_008762489.1 XP_008760711.1 7 1041
XM_008762490.1 XP_008760712.1 8 1098
XM_008762491.1 XP_008760713.1 7 1022
XM_008762492.1 XP_008760714.1 5 2313
XM_008762493.1 XP_008760715.1 7 1049
XM_008762494.1 XP_008760716.1 6 2532
GenBank mRNA BC083707.1 - 7 972

View Full Product Details