Gene Transcripts

Gene Symbol
Dnajc16
Entrez Gene ID
362652
Gene Name
DnaJ (Hsp40) homolog, subfamily C, member 16
Gene Aliases
Chromosome Location
Chr. 5: 160325088 - 160352874 on Build Rnor_6.0
UniGene ID
Rn.198307
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001014194.1 NP_001014216.1 4 488
GenBank mRNA BC089875.1 - 4 488

View Full Product Details