Gene Transcripts

Gene Symbol
Pdss2
Entrez Gene ID
365592
Gene Name
prenyl (decaprenyl) diphosphate synthase, subunit 2
Gene Aliases
Chromosome Location
Chr. 20: 47966511 - 48192747 on Build Rnor_6.0
UniGene ID
Rn.20063
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001014249.1 NP_001014271.1 8 1427
GenBank mRNA BC085898.1 - 8 1427

View Full Product Details