Gene Transcripts

Gene Symbol
C1orf53
Entrez Gene ID
388722
Gene Name
chromosome 1 open reading frame 53
Gene Aliases
Chromosome Location
Chr. 1: 197902552 - 197907367 on Build GRCh38
UniGene ID
Hs.61329
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001024594.2 NP_001019765.1 1 131
GenBank mRNA BC038214.1 - 1 30
BC130608.1 - 1 131 40146639
BC130634.1 - 1 131 40146665

View Full Product Details