Gene Transcripts

Gene Symbol
CATSPER4
Entrez Gene ID
378807
Gene Name
cation channel sperm associated 4
Gene Aliases
Chromosome Location
Chr. 1: 26189962 - 26202542 on Build GRCh38
UniGene ID
Hs.123532
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_198137.1 NP_937770.1 8 1121
XM_011541432.2 XP_011539734.1 8 1224
GenBank mRNA BC128138.1 - 8 1072
BC128139.1 - 8 1121 40112493
BC144663.1 - 7 1090

View Full Product Details