Gene Transcripts

Gene Symbol
Gata5
Entrez Gene ID
499951
Gene Name
GATA binding protein 5
Gene Aliases
Chromosome Location
Chr. 3: 175701278 - 175710852 on Build Rnor_6.0
UniGene ID
Rn.27175
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001024316.1 NP_001019487.1 6 1748
XM_008762533.1 XP_008760755.1 6 3213
GenBank mRNA BC085855.1 - 6 1748

View Full Product Details