Gene Transcripts

Gene Symbol
Lrrc73
Entrez Gene ID
501101
Gene Name
leucine rich repeat containing 73
Gene Aliases
nod3l
Chromosome Location
Chr. 9: 17111176 - 17115276 on Build Rnor_6.0
UniGene ID
Rn.51041
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001024360.2 NP_001019531.1 4 707
XM_008766878.1 XP_008765100.1 3 1683
GenBank mRNA AB210855.1 - 4 607
BC128712.1 - 4 707 7133504

View Full Product Details