Gene Transcripts

Gene Symbol
Cmklr1
Entrez Gene ID
14747
Gene Name
chemokine-like receptor 1
Gene Aliases
ChemR23, DEZ, Gpcr27, mcmklr1
Chromosome Location
Chr. 5: 113612355 - 113650450 on Build GRCm38
UniGene ID
Mm.5196
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_008153.3 NP_032179.1 3 983
GenBank mRNA AK158424.1 - 3 900
BC100612.1 - 3 983 2654794
U79525.1 - 3 986

View Full Product Details