Gene Transcripts

Gene Symbol
Mdga2
Entrez Gene ID
314180
Gene Name
MAM domain containing glycosylphosphatidylinositol anchor 2
Gene Aliases
Mamdc1
Chromosome Location
Chr. 6: 88446720 - 89292027 on Build Rnor_6.0
UniGene ID
Rn.205097
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_199269.1 NP_954890.1 5, 6 903
XM_008764688.1 XP_008762910.1 4, 5 1692
GenBank mRNA AY371924.1 - 5, 6 903

View Full Product Details