Gene Transcripts

Gene Symbol
ZCWPW1
Entrez Gene ID
55063
Gene Name
zinc finger CW-type and PWWP domain containing 1
Gene Aliases
ZCW1
Chromosome Location
Chr. 7: 100400826 - 100429239 on Build GRCh38
UniGene ID
Hs.105191
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001258008.1 NP_001244937.1 4 556
NM_017984.4 NP_060454.3 4 556
XM_005250479.2 XP_005250536.1 4 987
XM_005250480.2 XP_005250537.1 4 987
XM_006716035.3 XP_006716098.1 4 987
XM_006716036.2 XP_006716099.1 4 491
XM_006716037.3 XP_006716100.1 4 987
XM_006716038.3 XP_006716101.1 4 987
XM_006716040.3 XP_006716103.1 4 987
XM_017012379.1 XP_016867868.1 4 987
XM_017012380.1 XP_016867869.1 4 491
GenBank mRNA AK093038.1 - 4 491
AK299989.1 - 4 444
AK300745.1 - 4 412
AK302108.1 - 4 410
AK316553.1 - 4 744
AL136735.1 - 4 427
BC002725.1 - 1 145 3631266
DC413605.1 - 4 471

View Full Product Details