Gene Transcripts

Gene Symbol
FBXO42
Entrez Gene ID
54455
Gene Name
F-box protein 42
Gene Aliases
Fbx42, JFK
Chromosome Location
Chr. 1: 16246840 - 16352491 on Build GRCh38
UniGene ID
Hs.522384
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_018994.2 NP_061867.1 10 1784
XM_006710698.3 XP_006710761.1 9 1557
XM_017001502.1 XP_016856991.1 11 1913
XM_017001503.1 XP_016856992.1 11 1843
GenBank mRNA AB037753.1 - 9 1333
AK055598.1 - 10 1747
BC006174.1 - 6 918
BC043410.1 - 10 1746 6155685
BC063864.1 - 10 1746 6182628

View Full Product Details