Gene Transcripts

Gene Symbol
EQTN
Entrez Gene ID
54586
Gene Name
equatorin
Gene Aliases
AFAF, C9orf11, SPACA8
Chromosome Location
Chr. 9: 27284654 - 27297139 on Build GRCh38
UniGene ID
Hs.163070
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001161585.1 NP_001155057.1 5 467
NM_020641.2 NP_065692.2 6 554
XM_011517920.2 XP_011516222.1 3 1233
GenBank mRNA AJ278482.1 - 6 554
BC137351.1 - 6 524 9021349
BC137352.1 - 6 524 9021350
BC144602.1 - 5 437 9053133

View Full Product Details