Gene Transcripts

Gene Symbol
Pnma1
Entrez Gene ID
70481
Gene Name
paraneoplastic antigen MA1
Gene Aliases
5730402C15Rik
Chromosome Location
Chr. 12: 84146131 - 84148489 on Build GRCm38
UniGene ID
Mm.444348
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_027438.3 NP_081714.2 1 2145
GenBank mRNA AK017476.1 - 1 1939
AK028331.1 - 1 2083
AK136933.1 - 1 2147
BM939442.1 - 1 98

View Full Product Details