Gene Transcripts

Gene Symbol
Ttc8
Entrez Gene ID
76260
Gene Name
tetratricopeptide repeat domain 8
Gene Aliases
0610012F22Rik, AV001447, BBS8
Chromosome Location
Chr. 12: 98920574 - 98983250 on Build GRCm38
UniGene ID
Mm.282660
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_029553.3 NP_083829.1 13 1472
NM_198311.1 NP_938053.1 14 1464
GenBank mRNA AK002597.1 - 13 1458
AK032748.1 - 13 1434
AK081697.1 - 13 1439
BC017523.1 - 14 1464 4527657

View Full Product Details