Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_001012105.1 | NP_001012105.1 | 13 | 2058 | |
XM_006239303.1 | XP_006239365.1 | 13 | 2032 | ||
XM_008764253.1 | XP_008762475.1 | 12 | 1943 | ||
GenBank mRNA | BC083577.1 | - | 13 | 2058 |