Gene Transcripts

Gene Symbol
Rab34
Entrez Gene ID
360571
Gene Name
RAB34, member RAS oncogene family
Gene Aliases
Chromosome Location
Chr. 10: 65448111 - 65452178 on Build Rnor_6.0
UniGene ID
Rn.98493
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001012140.1 NP_001012140.1 7 631
GenBank mRNA BC086400.1 - 7 631

View Full Product Details