Gene Transcripts
- Gene Symbol
- Irf2bpl
- Entrez Gene ID
- 314329
- Gene Name
- interferon regulatory factor 2 binding protein-like
- Gene Aliases
- Eap1, Pqcp, RGD1310994
- Chromosome Location
- Chr. 6: 110901940 - 110904288 on Build Rnor_6.0
- UniGene ID
- Rn.15682
- Species
- Rattus norvegicus
View Full Product Details