Gene Transcripts

Gene Symbol
Ubxn10
Entrez Gene ID
298577
Gene Name
UBX domain protein 10
Gene Aliases
Ubxd3
Chromosome Location
Chr. 5: 157159931 - 157165767 on Build Rnor_6.0
UniGene ID
Rn.160610
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001013093.1 NP_001013111.1 2 916
GenBank mRNA BC079223.1 - 2 916

View Full Product Details