Gene Transcripts

Gene Symbol
ABCB8
Entrez Gene ID
11194
Gene Name
ATP binding cassette subfamily B member 8
Gene Aliases
EST328128, M-ABC1, MABC1
Chromosome Location
Chr. 7: 151028422 - 151047782 on Build GRCh38
UniGene ID
Hs.647118
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
GenBank mRNA AF047690.1 - 3 523

View Full Product Details