Gene Transcripts

Gene Symbol
SMCO3
Entrez Gene ID
440087
Gene Name
single-pass membrane protein with coiled-coil domains 3
Gene Aliases
C12orf69
Chromosome Location
Chr. 12: 14804650 - 14816761 on Build GRCh38
UniGene ID
Hs.220931
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001013698.2 NP_001013720.2 2 1430
XM_017019312.1 XP_016874801.1 3 4095
GenBank mRNA AK092637.1 - 2 1428
BC132752.1 - 2 1406 40146774
BC132754.1 - 2 1405 40146776

View Full Product Details