Gene Transcripts

Gene Symbol
Aspn
Entrez Gene ID
306805
Gene Name
asporin
Gene Aliases
Chromosome Location
Chr. 17: 14655958 - 14681356 on Build Rnor_6.0
UniGene ID
Rn.160633
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001014008.1 NP_001014030.1 814
XM_006253695.2 XP_006253757.1 2684
GenBank mRNA BC083711.1 - 814

Gene Symbol
NEWGENE_1549776
Entrez Gene ID
103690027
Gene Name
asporin
Gene Aliases
Aspn
Chromosome Location
Chr. 17: 15496470 - 15521144 on Build Rnor_6.0
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq XM_006253707.2 XP_006253769.1 6 2684
XM_008771557.1 XP_008769779.1 5 2760

View Full Product Details