Gene Transcripts
- Gene Symbol
- Tdrkh
- Entrez Gene ID
- 310652
- Gene Name
- tudor and KH domain containing
- Gene Aliases
- RGD1311196
- Chromosome Location
- Chr. 2: 195649835 - 195674217 on Build Rnor_6.0
- UniGene ID
- Rn.98848
- Species
- Rattus norvegicus
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
GenBank mRNA |
BC059161.1 |
- |
6 |
1149 |
|
View Full Product Details