Gene Transcripts

Gene Symbol
Dhrs11
Entrez Gene ID
360583
Gene Name
dehydrogenase/reductase (SDR family) member 11
Gene Aliases
RGD1307935
Chromosome Location
Chr. 10: 72144042 - 72153496 on Build Rnor_6.0
UniGene ID
Rn.204288
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001014119.2 NP_001014141.2 7 1365
XM_008768069.1 XP_008766291.1 6 1384
GenBank mRNA BC088444.1 - 7 1359
FQ233234.1 - 2 573

View Full Product Details