Gene Transcripts

Gene Symbol
Amz2
Entrez Gene ID
360650
Gene Name
archaelysin family metallopeptidase 2
Gene Aliases
RGD1304846
Chromosome Location
Chr. 10: 97710905 - 97721442 on Build Rnor_6.0
UniGene ID
Rn.15408
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001014121.1 NP_001014143.1 4 515
GenBank mRNA AJ879915.1 - 1 156
BC085886.1 - 4 515 7192275
FQ212663.1 - 4 457

View Full Product Details