Gene Transcripts

Gene Symbol
TMEM30B
Entrez Gene ID
161291
Gene Name
transmembrane protein 30B
Gene Aliases
CDC50B
Chromosome Location
Chr. 14: 61277370 - 61281812 on Build GRCh38
UniGene ID
Hs.146180
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001017970.2 NP_001017970.1 1 4027
GenBank mRNA AK091169.1 - 2 2928
AL080078.1 - 1 1778

View Full Product Details