Gene Transcripts

Gene Symbol
Tspan11
Entrez Gene ID
312727
Gene Name
tetraspanin 11
Gene Aliases
RGD1305424
Chromosome Location
Chr. 4: 160455848 - 160522905 on Build Rnor_6.0
UniGene ID
Rn.125922
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001024262.1 NP_001019433.1 8 1833
GenBank mRNA BC092652.1 - 8 1833

View Full Product Details