Gene Transcripts

Gene Symbol
Fam81a
Entrez Gene ID
76886
Gene Name
family with sequence similarity 81, member A
Gene Aliases
6430514L14Rik
Chromosome Location
Chr. 9: 70089167 - 70142555 on Build GRCm38
UniGene ID
Mm.44355
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_029784.2 NP_084060.1 9 2106
GenBank mRNA AK038532.1 - 9 2062
AK044090.1 - 9 2101
AK046408.1 - 9 1997
AK079541.1 - 9 2063

View Full Product Details