Gene Transcripts

Gene Symbol
Trappc6b
Entrez Gene ID
78232
Gene Name
trafficking protein particle complex 6B
Gene Aliases
5830498C14Rik, C79212
Chromosome Location
Chr. 12: 59043092 - 59061489 on Build GRCm38
UniGene ID
Mm.290070
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
GenBank mRNA AK020026.1 - 3 383
AK046610.1 - 3 411
AK089083.1 - 3 224
AK168294.1 - 4 671
BC031464.1 - 3 368 4017482

View Full Product Details