Gene Transcripts

Gene Symbol
Impact
Entrez Gene ID
497198
Gene Name
impact RWD domain protein
Gene Aliases
Chromosome Location
Chr. 18: 4325872 - 4346734 on Build Rnor_6.0
UniGene ID
Rn.190651
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001012235.1 NP_001012235.1 9, 10 838
GenBank mRNA AY574213.1 - 9, 10 838

View Full Product Details