Gene Transcripts

Gene Symbol
Izumo1
Entrez Gene ID
499152
Gene Name
izumo sperm-egg fusion 1
Gene Aliases
Chromosome Location
Chr. 1: 101603222 - 101608977 on Build Rnor_6.0
UniGene ID
Rn.125416
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001017514.1 NP_001017514.1 2 363
XM_008759406.1 XP_008757628.1 1 505
GenBank mRNA AB195683.1 - 1 127
BC079244.1 - 2 363 7132791

View Full Product Details