Gene Transcripts

Gene Symbol
Retnlb
Entrez Gene ID
498074
Gene Name
resistin like beta
Gene Aliases
Chromosome Location
Chr. 11: 54565471 - 54567357 on Build Rnor_6.0
UniGene ID
Rn.145442
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001024281.1 NP_001019452.1 4 422
GenBank mRNA AY641429.1 - 4 422

View Full Product Details