Gene Transcripts

Gene Symbol
Slc35d3
Entrez Gene ID
76157
Gene Name
solute carrier family 35, member D3
Gene Aliases
6230421J19Rik, Frcl1
Chromosome Location
Chr. 10: 19847917 - 19852361 on Build GRCm38
UniGene ID
Mm.74324
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_029529.3 NP_083805.1 2 2561
GenBank mRNA AK042761.1 - 2 2313
AK044973.1 - 2 2298
AK047610.1 - 2 2562

View Full Product Details