Gene Transcripts
- Gene Symbol
- TAP1
- Entrez Gene ID
- 6890
- Gene Name
- transporter 1, ATP binding cassette subfamily B member
- Gene Aliases
- ABC17, ABCB2, APT1, D6S114E, PSF-1, PSF1, RING4, TAP1*0102N, TAP1N
- Chromosome Location
- Chr. 6: 32845209 - 32853971 on Build GRCh38
- UniGene ID
- Hs.352018
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NM_000593.5 |
NP_000584.2
|
3, 4 |
1187 |
|
NM_001292022.1 |
NP_001278951.1
|
3, 4 |
447 |
|
GenBank mRNA |
AB012644.1 |
- |
3, 4 |
851 |
|
AK302340.1 |
- |
3, 4 |
418 |
|
AK313762.1 |
- |
3, 4 |
1223 |
|
AY523970.1 |
- |
3, 4 |
1058 |
|
AY523971.2 |
- |
3, 4 |
1058 |
|
BC014081.2 |
- |
3, 4 |
1187 |
4304716 |
BX648013.1 |
- |
3, 4 |
1195 |
|
DC354917.1 |
- |
3, 4 |
447 |
|
KU178346.1 |
- |
3, 4 |
1032 |
|
L21204.1 |
- |
3, 4 |
852 |
|
L21205.1 |
- |
3, 4 |
852 |
|
L21206.1 |
- |
3, 4 |
852 |
|
L21207.1 |
- |
3, 4 |
852 |
|
L21208.1 |
- |
3, 4 |
852 |
|
X57521.1 |
- |
3, 4 |
486 |
|
X57522.1 |
- |
3, 4 |
1062 |
|
View Full Product Details