Gene Transcripts

Gene Symbol
PLIN4
Entrez Gene ID
729359
Gene Name
perilipin 4
Gene Aliases
KIAA1881, S3-12
Chromosome Location
Chr. 19: 4502180 - 4520285 on Build GRCh38
UniGene ID
Hs.591387
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001080400.1 NP_001073869.1 6 5656
XM_006722866.2 XP_006722929.1 8 7617
XM_006722868.3 XP_006722931.1 8 7613
XM_011528233.2 XP_011526535.1 9 7799
XM_017027192.1 XP_016882681.1 9 7802
XM_017027193.1 XP_016882682.1 9 7749
XM_017027194.1 XP_016882683.1 9 7784
GenBank mRNA AB067468.1 - 5630

View Full Product Details