Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NR_134984.1 | 1 | 638 | ||
NR_134985.1 | 1 | 638 | |||
NR_134986.1 | 1 | 638 | |||
NR_134987.1 | 1 | 638 | |||
GenBank mRNA | AK026760.1 | - | 1 | 21 | |
AK124047.1 | - | 1 | 638 | ||
CB152000.1 | - | 1 | 534 |