Gene Transcripts

Gene Symbol
VRTN
Entrez Gene ID
55237
Gene Name
vertebrae development associated
Gene Aliases
C14orf115, vertnin
Chromosome Location
Chr. 14: 74303076 - 74360008 on Build GRCh38
UniGene ID
Hs.578167
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_018228.2 NP_060698.2 2 562
XM_011536911.1 XP_011535213.1 3 609
GenBank mRNA AK001673.1 - 2 516
AK023213.1 - 2 529
BC053325.1 - 2 545 6577658
CX165172.1 - 2 562

View Full Product Details