Gene Transcripts

Gene Symbol
MSL2
Entrez Gene ID
55167
Gene Name
male-specific lethal 2 homolog (Drosophila)
Gene Aliases
MSL-2, MSL2L1, RNF184
Chromosome Location
Chr. 3: 136148918 - 136197300 on Build GRCh38
UniGene ID
Hs.18631
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001145417.1 NP_001138889.1 2 2726
NM_018133.3 NP_060603.2 2 3527
XM_005247571.3 XP_005247628.1 2 3261
XM_006713684.3 XP_006713747.1 2 2744
XM_011512949.2 XP_011511251.1 2 2831
GenBank mRNA AB046805.1 - 2 2875
AK001408.1 - 1 1780
AL834289.1 - 2 2845
BC032719.1 - 1 2316

View Full Product Details