Gene Transcripts
- Gene Symbol
- TTC41P
- Entrez Gene ID
- 253724
- Gene Name
- tetratricopeptide repeat domain 41, pseudogene
- Gene Aliases
- GNN, GNNP
- Chromosome Location
- Chr. 12: 103843749 - 103930211 on Build GRCh38
- UniGene ID
- Hs.506549
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NR_027249.1 |
|
2 |
1205 |
|
GenBank mRNA |
BC016335.1 |
- |
2 |
1037 |
|
BC033128.1 |
- |
6 |
1786 |
|
View Full Product Details