Gene Transcripts

Gene Symbol
C11orf86
Entrez Gene ID
254439
Gene Name
chromosome 11 open reading frame 86
Gene Aliases
Chromosome Location
Chr. 11: 66975283 - 66977008 on Build GRCh38
UniGene ID
Hs.232604
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001136485.1 NP_001129957.1 2 636
GenBank mRNA AK026328.1 - 2 642

View Full Product Details