Gene Transcripts

Gene Symbol
DLX6-AS1
Entrez Gene ID
285987
Gene Name
DLX6 antisense RNA 1
Gene Aliases
DLX6-AS, DLX6AS, Evf-2, NCRNA00212
Chromosome Location
Chr. 7: 96968515 - 97014065 on Build GRCh38
UniGene ID
Hs.34969
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
GenBank mRNA AK055423.1 - 1 263
AK091367.1 - 2 2522
AK095619.1 - 3 3117
AK095764.1 - 1 1320

View Full Product Details