Gene Transcripts

Gene Symbol
SUPT20HL2
Entrez Gene ID
170067
Gene Name
SPT20 homolog, SAGA complex component-like 2
Gene Aliases
FAM48B1, FAM48B2
Chromosome Location
Chr. X: 24310862 - 24313315 on Build GRCh38
UniGene ID
Hs.716893
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001136233.1 NP_001129705.1 1 2342

Gene Symbol
SUPT20HL1
Entrez Gene ID
100130302
Gene Name
SPT20 homolog, SAGA complex component-like 1
Gene Aliases
FAM48B1, SPT20L
Chromosome Location
Chr. X: 24362761 - 24365424 on Build GRCh38
UniGene ID
Hs.717243
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001136234.1 NP_001129706.1 1 2552

View Full Product Details