Gene Transcripts
- Gene Symbol
- SEPT11
- Entrez Gene ID
- 55752
- Gene Name
- septin 11
- Gene Aliases
- Chromosome Location
- Chr. 4: 76949714 - 77040154 on Build GRCh38
- UniGene ID
- Hs.128199
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NM_001306147.1 |
NP_001293076.1
|
11, 12 |
1681 |
|
NM_018243.3 |
NP_060713.1
|
9, 10 |
1461 |
|
XM_006714259.3 |
XP_006714322.1
|
9, 10 |
1327 |
|
XM_017008400.1 |
XP_016863889.1
|
10, 11 |
1470 |
|
GenBank mRNA |
AK001711.1 |
- |
9, 10 |
1364 |
|
AK027279.1 |
- |
7, 8 |
1038 |
|
AK027633.1 |
- |
6, 7 |
876 |
|
AK302700.1 |
- |
11, 12 |
1558 |
|
BC008083.2 |
- |
9, 10 |
1447 |
2966709 |
BC063615.1 |
- |
9, 10 |
1312 |
3848367 |
GU727629.1 |
- |
9, 10 |
1273 |
|
View Full Product Details