Gene Transcripts

Gene Symbol
SRCIN1
Entrez Gene ID
80725
Gene Name
SRC kinase signaling inhibitor 1
Gene Aliases
P140, SNIP
Chromosome Location
Chr. 17: 38530015 - 38607166 on Build GRCh38
UniGene ID
Hs.448872
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_025248.2 NP_079524.2 19 6883
XM_017025169.1 XP_016880658.1 23 7975
XM_017025170.1 XP_016880659.1 21 7173
XM_017025171.1 XP_016880660.1 20 7050
XM_017025172.1 XP_016880661.1 19 6948
GenBank mRNA BC019717.1 - 1 621

View Full Product Details