Gene Transcripts
- Gene Symbol
- CISD1
- Entrez Gene ID
- 55847
- Gene Name
- CDGSH iron sulfur domain 1
- Gene Aliases
- C10orf70, MDS029, ZCD1, mitoNEET
- Chromosome Location
- Chr. 10: 58269102 - 58290553 on Build GRCh38
- UniGene ID
- Hs.370102
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NM_018464.4 |
NP_060934.1
|
2, 3 |
407 |
|
XM_017016409.1 |
XP_016871898.1
|
3, 4 |
648 |
|
GenBank mRNA |
AF220049.1 |
- |
2, 3 |
346 |
|
AK312017.1 |
- |
2, 3 |
282 |
|
AY960578.1 |
- |
2, 3 |
290 |
|
BC005962.1 |
- |
2, 3 |
383 |
4051044 |
BC007043.1 |
- |
2, 3 |
339 |
3930701 |
BC008474.1 |
- |
2, 3 |
344 |
4283253 |
BC059168.1 |
- |
2, 3 |
354 |
4132410 |
DA417986.1 |
- |
2, 3 |
407 |
|
View Full Product Details