Gene Transcripts

Gene Symbol
CAPN14
Entrez Gene ID
440854
Gene Name
calpain 14
Gene Aliases
Chromosome Location
Chr. 2: 31173056 - 31233957 on Build GRCh38
UniGene ID
Hs.468059
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001145122.1 NP_001138594.1 10 1106
NM_001321270.1 NP_001308199.1 10 1170
XM_011532863.2 XP_011531165.1 11 1267
XM_011532864.2 XP_011531166.1 11 1304
XM_011532865.1 XP_011531167.1 11 1208
XM_011532866.2 XP_011531168.1 9 975
GenBank mRNA AK092257.1 - 10 1071
AK296910.1 - 12 1074

View Full Product Details