Gene Transcripts
- Gene Symbol
- COL6A4P2
- Entrez Gene ID
- 646300
- Gene Name
- collagen type VI alpha 4 pseudogene 2
- Gene Aliases
- COL6A4
- Chromosome Location
- Chr. 3: 130212820 - 130273806 on Build GRCh38
- UniGene ID
- Hs.170475
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NR_027898.1 |
|
12 |
639 |
|
GenBank mRNA |
CN285077.1 |
- |
10 |
543 |
|
View Full Product Details